Gene Transcripts

Gene Symbol
Olr40
Entrez Gene ID
293197
Gene Name
olfactory receptor 40
Gene Aliases
ratchr1-160170992-160170042_ORF
Chromosome Location
Chr. 1: 167605393 - 167606343 on Build Rnor_6.0
UniGene ID
Rn.141934
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000127.1 NP_001000127.1 1 578

View Full Product Details