Gene Transcripts

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
Chromosome Location
Chr. 6: 169751622 - 169781584 on Build GRCh38
UniGene ID
Hs.47546
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278531.1 NP_001265460.1 14, 15 1431
NM_001278532.1 NP_001265461.1 13, 14 1262
NM_001278533.1 NP_001265462.1 14, 15 1431
NM_018341.2 NP_060811.1 14, 15 1431
XM_011535938.2 XP_011534240.1 13, 14 1401
XM_011535939.2 XP_011534241.1 14, 15 2521
XM_011535940.2 XP_011534242.1 14, 15 2330
XM_017011027.1 XP_016866516.1 14, 15 2271
XM_017011028.1 XP_016866517.1 14, 15 1596
XM_017011029.1 XP_016866518.1 14, 15 1356
XM_017011030.1 XP_016866519.1 13, 14 1180
XM_017011031.1 XP_016866520.1 13, 14 1245
XM_017011032.1 XP_016866521.1 14, 15 2516
XM_017011033.1 XP_016866522.1 14, 15 2325
GenBank mRNA AK001369.1 - 13, 14 1292
AK002014.1 - 14, 15 1428
AK021520.1 - 4, 5 1495
AK294090.1 - 13, 14 1259
BC044812.1 - 14, 15 1415

View Full Product Details