Gene Transcripts

Gene Symbol
Treml1
Entrez Gene ID
71326
Gene Name
triggering receptor expressed on myeloid cells-like 1
Gene Aliases
5430401J17Rik, TLT-1
Chromosome Location
Chr. 17: 48359916 - 48367176 on Build GRCm38
UniGene ID
Mm.90177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027763.2 NP_082039.1 3, 4 547
GenBank mRNA AY078502.1 - 3, 4 547

View Full Product Details