Gene Transcripts

Gene Symbol
Fhad1
Entrez Gene ID
329977
Gene Name
forkhead-associated (FHA) phosphopeptide binding domain 1
Gene Aliases
2900090M10Rik, B230311F11
Chromosome Location
Chr. 4: 141890430 - 142015124 on Build GRCm38
UniGene ID
Mm.252019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177868.4 NP_808536.2 20, 21 2827
GenBank mRNA AK148490.1 - 5, 6 912
BC038692.1 - 2, 3 410 1496088

View Full Product Details