Gene Transcripts

Gene Symbol
TTC38
Entrez Gene ID
55020
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
LL22NC03-5H6.5
Chromosome Location
Chr. 22: 46267964 - 46294008 on Build GRCh38
UniGene ID
Hs.435832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017931.2 NP_060401.2 14 2577
XM_011530259.1 XP_011528561.1 15 2705
GenBank mRNA AK000706.1 - 14 2531
BC018918.2 - 14 2518 4130906

View Full Product Details