Gene Transcripts

Gene Symbol
NOL8
Entrez Gene ID
55035
Gene Name
nucleolar protein 8
Gene Aliases
C9orf34, NOP132, bA62C3.3, bA62C3.4
Chromosome Location
Chr. 9: 92297358 - 92325594 on Build GRCh38
UniGene ID
Hs.442199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256394.1 NP_001243323.1 8 1628
NM_017948.5 NP_060418.4 7 1524
NR_046106.1 7 1524
XM_006717166.3 XP_006717229.1 7 1390
XM_006717167.3 XP_006717230.1 7 1512
XM_006717168.3 XP_006717231.1 7 1283
XM_006717169.3 XP_006717232.1 8 1543
XM_006717170.3 XP_006717233.1 8 1411
XM_006717172.3 XP_006717235.1 8 1735
XM_006717173.3 XP_006717236.1 9 1848
XM_011518824.2 XP_011517126.1 6 1108
XM_011518827.2 XP_011517129.1 5 1152
XM_017014876.1 XP_016870365.1 7 1390
XM_017014877.1 XP_016870366.1 8 1411
XM_017014878.1 XP_016870367.1 8 1861
XM_017014879.1 XP_016870368.1 8 1690
XM_017014880.1 XP_016870369.1 8 1490
GenBank mRNA AB105104.1 - 7 1490
AB109030.1 - 6 1187
AK001049.1 - 7 1406
AK022755.1 - 4 937
AK024245.1 - 1 282
AK297355.1 - 7 1407
AK307408.1 - 7 1415
BC064942.1 - 1 447
BC146810.1 - 8 1384 8860369
CR627363.1 - 7 1268

View Full Product Details