Gene Transcripts

Gene Symbol
Rab18
Entrez Gene ID
307039
Gene Name
RAB18, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 17: 59844724 - 59876722 on Build Rnor_6.0
UniGene ID
Rn.221766
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012468.1 NP_001012486.1 7 1194
XM_006254132.2 XP_006254194.1 7 1251
GenBank mRNA BC089957.1 - 7 1194

View Full Product Details