Gene Transcripts

Gene Symbol
Rmdn3
Entrez Gene ID
311328
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
Fam82a2, Fam82c, Ptpip51, RGD1308697, Rmd-3
Chromosome Location
Chr. 3: 110943843 - 110964449 on Build Rnor_6.0
UniGene ID
Rn.18772
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014046.2 NP_001014068.1 4, 5 618
GenBank mRNA BC082081.1 - 4, 5 580
DN931790.1 - 4, 5 650

View Full Product Details