Gene Transcripts

Gene Symbol
Abcg3l2
Entrez Gene ID
360910
Gene Name
ATP-binding cassette, subfamily G (WHITE), member 3-like 2
Gene Aliases
Chromosome Location
Chr. 14: 5568023 - 5609333 on Build Rnor_6.0
UniGene ID
Rn.36147
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014133.1 NP_001014155.1 1115
GenBank mRNA BC085773.1 - 1115

View Full Product Details