Gene Transcripts

Gene Symbol
Fli1
Entrez Gene ID
315532
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Gene Aliases
Chromosome Location
Chr. 8: 33541932 - 33661094 on Build Rnor_6.0
UniGene ID
Rn.203677
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017381.1 NP_001017381.1 5 919
XM_008766055.1 XP_008764277.1 5 954
GenBank mRNA AY972521.1 - 5 919

View Full Product Details