Gene Transcripts

Gene Symbol
Kdm6b
Entrez Gene ID
216850
Gene Name
KDM1 lysine (K)-specific demethylase 6B
Gene Aliases
1700064E03Rik, BC038313, Jmjd3, mKIAA0346
Chromosome Location
Chr. 11: 69398508 - 69418148 on Build GRCm38
UniGene ID
Mm.261201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017426.1 NP_001017426.1 20 4995
GenBank mRNA AK129121.1 - 10 1787
AK136019.1 - 10 1176
BC029201.1 - 1 89
BC038313.1 - 5 633
BC075632.1 - 21 5171 30362899

View Full Product Details