Gene Transcripts

Gene Symbol
Gtpbp8
Entrez Gene ID
360714
Gene Name
GTP-binding protein 8 (putative)
Gene Aliases
RGD1310025
Chromosome Location
Chr. 11: 60906990 - 60916958 on Build Rnor_6.0
UniGene ID
Rn.138748
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025015.1 NP_001020186.1 5 780
XM_006248340.2 XP_006248402.1 4 706
GenBank mRNA BC091236.1 - 5 780
FQ216550.1 - 5 779

View Full Product Details