Gene Transcripts

Gene Symbol
Mup3
Entrez Gene ID
17842
Gene Name
major urinary protein 3
Gene Aliases
MUP15, MUPIII, Mup-3, Mup25
Chromosome Location
Chr. 4: 62083476 - 62087370 on Build GRCm38
UniGene ID
Mm.250267
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA M27608.1 - 6, 7 732

View Full Product Details