Gene Transcripts

Gene Symbol
WDR70
Entrez Gene ID
55100
Gene Name
WD repeat domain 70
Gene Aliases
Chromosome Location
Chr. 5: 37379280 - 37752672 on Build GRCh38
UniGene ID
Hs.213690
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018034.2 NP_060504.1 5 471
XM_005248318.3 XP_005248375.1 5 489
GenBank mRNA AK001095.1 - 5 413
AK291179.1 - 5 493
AK310901.1 - 5 496
AL512685.1 - 5 472
BC009648.1 - 5 471 3454326
BC025315.1 - 5 449 5458850
BX537655.1 - 4 428

View Full Product Details