Gene Transcripts
- Gene Symbol
- ANO10
- Entrez Gene ID
- 55129
- Gene Name
- anoctamin 10
- Gene Aliases
- SCAR10, TMEM16K
- Chromosome Location
- Chr. 3: 43366324 - 43691594 on Build GRCh38
- UniGene ID
- Hs.656657
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204831.1 |
NP_001191760.1
|
4, 5 |
639 |
|
NM_001204832.1 |
NP_001191761.1
|
3, 4 |
441 |
|
NM_001204834.1 |
NP_001191763.1
|
4, 5 |
639 |
|
NM_018075.3 |
NP_060545.3
|
4, 5 |
639 |
|
XM_011533885.2 |
XP_011532187.2
|
4, 5 |
605 |
|
XM_011533887.2 |
XP_011532189.1
|
4, 5 |
605 |
|
XM_011533889.2 |
XP_011532191.1
|
4, 5 |
605 |
|
XM_011533890.2 |
XP_011532192.1
|
4, 5 |
605 |
|
XM_017006716.1 |
XP_016862205.1
|
4, 5 |
605 |
|
XM_017006717.1 |
XP_016862206.1
|
4, 5 |
557 |
|
XM_017006718.1 |
XP_016862207.1
|
4, 5 |
584 |
|
XM_017006719.1 |
XP_016862208.1
|
3, 4 |
359 |
|
XM_017006720.1 |
XP_016862209.1
|
4, 5 |
584 |
|
XM_017006721.1 |
XP_016862210.1
|
3, 4 |
368 |
|
XM_017006722.1 |
XP_016862211.1
|
3, 4 |
341 |
|
GenBank mRNA |
AK096302.1 |
- |
4, 5 |
639 |
|
AK131223.1 |
- |
3, 4 |
357 |
|
AK292368.1 |
- |
4, 5 |
524 |
|
AK297949.1 |
- |
4, 5 |
582 |
|
AL832508.1 |
- |
4, 5 |
506 |
|
BC038855.1 |
- |
4, 5 |
537 |
|
View Full Product Details