Gene Transcripts

Gene Symbol
OXR1
Entrez Gene ID
55074
Gene Name
oxidation resistance 1
Gene Aliases
Nbla00307, TLDC3
Chromosome Location
Chr. 8: 106270178 - 106752694 on Build GRCh38
UniGene ID
Hs.127286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198532.1 NP_001185461.1 16 3437
NM_001198533.1 NP_001185462.1 17 3663
NM_001198534.1 NP_001185463.1 7 1582
NM_001198535.1 NP_001185464.1 6 1501
NM_018002.3 NP_060472.2 16 3582
NM_181354.4 NP_851999.2 14 3412
XM_006716595.2 XP_006716658.1 15 3351
XM_017013589.1 XP_016869078.1 15 3462
XM_017013590.1 XP_016869079.1 15 3303
XM_017013591.1 XP_016869080.1 15 3393
XM_017013592.1 XP_016869081.1 15 3140
XM_017013593.1 XP_016869082.1 15 3367
XM_017013594.1 XP_016869083.1 14 3286
XM_017013595.1 XP_016869084.1 14 3277
XM_017013596.1 XP_016869085.1 16 3450
XM_017013597.1 XP_016869086.1 14 3111
XM_017013598.1 XP_016869087.1 14 3182
XM_017013599.1 XP_016869088.1 13 3196
XM_017013600.1 XP_016869089.1 13 3053
GenBank mRNA AB075503.1 - 7 1527
AK000987.1 - 7 1553
AK098168.1 - 1 1927
AK123667.1 - 6 1816
AL833193.1 - 13 2992
FN650108.1 - 16 3473

View Full Product Details