Gene Transcripts

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
Chromosome Location
Chr. 11: 70078302 - 70189545 on Build GRCh38
UniGene ID
Hs.503074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018043.5 NP_060513.5 26 4193
XM_006718602.2 XP_006718665.1 27 3974
XM_006718604.2 XP_006718667.1 27 4089
XM_006718605.2 XP_006718668.1 24 3945
XM_011545121.2 XP_011543423.1 27 3966
XM_011545123.2 XP_011543425.1 27 3963
XM_011545124.2 XP_011543426.1 26 3954
XM_011545125.2 XP_011543427.1 26 3888
XM_011545126.2 XP_011543428.1 25 3876
XM_011545127.2 XP_011543429.1 25 3822
XM_011545128.2 XP_011543430.1 24 3810
XM_011545129.2 XP_011543431.1 27 3881
XM_017017956.1 XP_016873445.1 26 3897
XM_017017957.1 XP_016873446.1 25 3888
GenBank mRNA AK001123.1 - 16 2731
AL833582.1 - 26 3937
AY728143.1 - 26 4193
BC027590.2 - 20 3067

View Full Product Details