Gene Transcripts

Gene Symbol
Mrgpra4
Entrez Gene ID
235854
Gene Name
MAS-related GPR, member A4
Gene Aliases
Mgrg10, MrgA4
Chromosome Location
Chr. 7: 47980794 - 47982296 on Build GRCm38
UniGene ID
Mm.207065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153524.2 NP_705744.2 1 1480
GenBank mRNA AY042194.1 - 1 1480

View Full Product Details