Gene Transcripts

Gene Symbol
PROX2
Entrez Gene ID
283571
Gene Name
prospero homeobox 2
Gene Aliases
PROX-2
Chromosome Location
Chr. 14: 74853033 - 74876663 on Build GRCh38
UniGene ID
Hs.675293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080408.2 NP_001073877.2 1 69
NM_001243007.1 NP_001229936.1 1 69
XM_005267543.3 XP_005267600.1 3 1147
XM_017021222.1 XP_016876711.1 2 1012
GenBank mRNA BC105720.1 - 1 68
BC105927.1 - 1 68

View Full Product Details