Gene Transcripts

Gene Symbol
METTL11B
Entrez Gene ID
149281
Gene Name
methyltransferase like 11B
Gene Aliases
C1orf184, HOMT1B, NTM1B
Chromosome Location
Chr. 1: 170146047 - 170168841 on Build GRCh38
UniGene ID
Hs.553612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136107.1 NP_001129579.1 1 166
XM_011509232.2 XP_011507534.1 1 38
GenBank mRNA BC157860.1 - 1 166
BC171858.1 - 1 166 9054512

View Full Product Details