Gene Transcripts

Gene Symbol
SHISA3
Entrez Gene ID
152573
Gene Name
shisa family member 3
Gene Aliases
hShisa3
Chromosome Location
Chr. 4: 42397839 - 42402487 on Build GRCh38
UniGene ID
Hs.370904
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080505.1 NP_001073974.1 2 1204
GenBank mRNA BC012029.1 - 1 704

View Full Product Details