Gene Transcripts

Gene Symbol
R3HCC1
Entrez Gene ID
203069
Gene Name
R3H domain and coiled-coil containing 1
Gene Aliases
Chromosome Location
Chr. 8: 23288092 - 23296279 on Build GRCh38
UniGene ID
Hs.458644
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC143822.1 - 1 42
BC143827.1 - 1 42
BC143829.1 - 1 42 9052344

View Full Product Details