Gene Transcripts

Gene Symbol
MSTO1
Entrez Gene ID
55154
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
LST005, MST
Chromosome Location
Chr. 1: 155610170 - 155614967 on Build GRCh38
UniGene ID
Hs.656547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256532.1 NP_001243461.1 7 696
NM_001256533.1 NP_001243462.1 7 696
NM_018116.3 NP_060586.2 7 696
NR_046292.1 7 818
NR_046293.1 7 760
NR_046294.1 5 682
NR_046295.1 6 620
XM_006711421.1 XP_006711484.1 6 655
XM_006711423.2 XP_006711486.1 7 704
XM_011509695.1 XP_011507997.1 7 654
XM_011509696.1 XP_011507998.1 7 654
XM_011509697.1 XP_011507999.1 7 654
XM_011509698.1 XP_011508000.1 7 654
XM_011509699.1 XP_011508001.1 7 654
XM_011509700.1 XP_011508002.1 7 654
XM_011509701.1 XP_011508003.1 7 654
XM_011509702.1 XP_011508004.1 7 654
XM_011509703.1 XP_011508005.1 6 1296
XM_011509704.1 XP_011508006.1 7 1219
XM_011509705.1 XP_011508007.1 6 1233
XM_011509706.1 XP_011508008.1 7 653
XM_011509707.1 XP_011508009.1 7 1161
XM_011509708.1 XP_011508010.1 7 1097
XM_011509709.2 XP_011508011.1 7 654
XM_017001603.1 XP_016857092.1 7 717
XM_017001604.1 XP_016857093.1 7 655
XM_017001605.1 XP_016857094.1 7 1186
XM_017001606.1 XP_016857095.1 7 1097
XM_017001607.1 XP_016857096.1 7 1235
XM_017001608.1 XP_016857097.1 6 682
XM_017001609.1 XP_016857098.1 7 704
XM_017001610.1 XP_016857099.1 7 692
XM_017001611.1 XP_016857100.1 7 1223
XM_017001612.1 XP_016857101.1 6 597
XM_017001613.1 XP_016857102.1 6 595
XM_017001614.1 XP_016857103.1 7 704
XM_017001615.1 XP_016857104.1 7 766
XM_017001616.1 XP_016857105.1 3 398
GenBank mRNA AF111708.1 - 4 385
AF272833.1 - 7 574
AK001366.1 - 7 625
AK056128.1 - 5 611
AK092950.1 - 6 520
AK222863.1 - 7 622
AK290610.1 - 7 671
AK297140.1 - 7 747
AK309573.1 - 7 681
AY334564.1 - 6 739
BC002535.2 - 7 641 3140498
BC070067.1 - 7 712
BX537684.1 - 6 572
CR749791.1 - 7 622
CR936872.1 - 3 1664

Gene Symbol
MSTO2P
Entrez Gene ID
100129405
Gene Name
misato family member 2, pseudogene
Gene Aliases
MSTO2
Chromosome Location
Chr. 1: 155745768 - 155750688 on Build GRCh38
UniGene ID
Hs.656361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024117.2 7 742
GenBank mRNA AK293928.1 - 7 678
AK295146.1 - 7 742
BC099909.1 - 7 692

View Full Product Details