Gene Transcripts

Gene Symbol
ZFR2
Entrez Gene ID
23217
Gene Name
zinc finger RNA binding protein 2
Gene Aliases
KIAA1086
Chromosome Location
Chr. 19: 3804024 - 3869029 on Build GRCh38
UniGene ID
Hs.65750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015174.1 NP_055989.1 6, 7 1046
XM_011527831.2 XP_011526133.1 6, 7 1045
GenBank mRNA AB029009.1 - 5, 6 924

View Full Product Details