Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015325.2 | NP_056140.1 | 13 | 2253 | |
XM_011513999.2 | XP_011512301.1 | 13 | 2253 | ||
XM_017009285.1 | XP_016864774.1 | 14 | 2342 | ||
GenBank mRNA | AB023164.3 | - | 13 | 2232 | |
AK096615.1 | - | 1 | 483 | ||
AL832204.1 | - | 1 | 724 | ||
CR749441.1 | - | 6 | 1524 |