Gene Transcripts

Gene Symbol
KIAA0368
Entrez Gene ID
23392
Gene Name
KIAA0368
Gene Aliases
ECM29
Chromosome Location
Chr. 9: 111360693 - 111484745 on Build GRCh38
UniGene ID
Hs.368255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080398.1 NP_001073867.1 16, 17 1997
XM_005251850.3 XP_005251907.1 14, 15 1500
XM_005251851.4 XP_005251908.1 15, 16 1879
XM_005251852.4 XP_005251909.1 15, 16 1831
XM_005251853.3 XP_005251910.1 15, 16 1613
XM_005251854.3 XP_005251911.1 14, 15 1782
XM_011518422.2 XP_011516724.1 14, 15 1500
XM_017014557.1 XP_016870046.1 8, 9 819
GenBank mRNA AB002366.1 - 4, 5 271
AK122712.1 - 14, 15 1484
AK127247.1 - 10, 11 940
AK308313.1 - 15, 16 1604

View Full Product Details