Gene Transcripts

Gene Symbol
SLC22A10
Entrez Gene ID
387775
Gene Name
solute carrier family 22 member 10
Gene Aliases
OAT5, hOAT5
Chromosome Location
Chr. 11: 63268257 - 63311983 on Build GRCh38
UniGene ID
Hs.188982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039752.3 NP_001034841.3 10 1866
NR_134874.1 11 1482
XM_011545016.2 XP_011543318.1 12 2871
XM_017017699.1 XP_016873188.1 11 1668
XM_017017700.1 XP_016873189.1 10 2104
XM_017017702.1 XP_016873191.1 9 2343
XM_017017703.1 XP_016873192.1 9 1212
GenBank mRNA AB075876.1 - 11 1482
AK298430.1 - 11 1529

View Full Product Details