Gene Transcripts

Gene Symbol
C17orf98
Entrez Gene ID
388381
Gene Name
chromosome 17 open reading frame 98
Gene Aliases
Chromosome Location
Chr. 17: 38835088 - 38841389 on Build GRCh38
UniGene ID
Hs.250182
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080465.2 NP_001073934.1 2 276
GenBank mRNA DY654789.1 - 2 227

View Full Product Details