Gene Transcripts
- Gene Symbol
- CYP4F24P
- Entrez Gene ID
- 388514
- Gene Name
- cytochrome P450 family 4 subfamily F member 24, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 19: 15759334 - 15779988 on Build GRCh38
- UniGene ID
- Hs.438274
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033864.1 |
|
2 |
301 |
|
GenBank mRNA |
AK297837.1 |
- |
3 |
449 |
|
AK308750.1 |
- |
2 |
296 |
|
View Full Product Details