Gene Transcripts

Gene Symbol
ANKUB1
Entrez Gene ID
389161
Gene Name
ankyrin repeat and ubiquitin domain containing 1
Gene Aliases
C3orf16
Chromosome Location
Chr. 3: 149760648 - 149792823 on Build GRCh38
UniGene ID
Hs.665988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144960.2 NP_001138432.1 5 1165
NM_001315505.1 NP_001302434.1 5 1055
NM_001315506.1 NP_001302435.1 5 1165
XM_011512798.1 XP_011511100.1 5 662
XM_011512799.1 XP_011511101.1 5 896
XM_011512801.1 XP_011511103.1 3 333
GenBank mRNA AK027233.1 - 2 164
AK304359.1 - 5 896
BC171805.1 - 5 1055

View Full Product Details