Gene Transcripts
- Gene Symbol
- EGFEM1P
- Entrez Gene ID
- 93556
- Gene Name
- EGF like and EMI domain containing 1, pseudogene
- Gene Aliases
- C3orf50, NCRNA00259
- Chromosome Location
- Chr. 3: 168249522 - 168830599 on Build GRCh38
- UniGene ID
- Hs.478158
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_021485.2 |
|
2 |
596 |
|
GenBank mRNA |
BI461452.1 |
- |
2 |
458 |
|
DR000794.1 |
- |
2 |
603 |
|
View Full Product Details