Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002612.3 | NP_002603.1 | 2 | 538 | |
GenBank mRNA | AK096428.1 | - | 3 | 337 | |
AK223328.1 | - | 2 | 434 | ||
AK312424.1 | - | 2 | 444 | ||
BC040239.1 | - | 2 | 418 | 3047987 | |
DB035430.1 | - | 2 | 540 | ||
U54617.1 | - | 2 | 440 |