Gene Transcripts
- Gene Symbol
- TEX2
- Entrez Gene ID
- 55852
- Gene Name
- testis expressed 2
- Gene Aliases
- HT008, TMEM96
- Chromosome Location
- Chr. 17: 64147433 - 64263323 on Build GRCh38
- UniGene ID
- Hs.175414
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288732.1 |
NP_001275661.1
|
9, 10 |
3118 |
|
NM_001288733.1 |
NP_001275662.1
|
9, 10 |
3110 |
|
NM_018469.4 |
NP_060939.3
|
9, 10 |
3139 |
|
XM_011524998.1 |
XP_011523300.1
|
9, 10 |
3128 |
|
XM_011524999.1 |
XP_011523301.1
|
9, 10 |
3052 |
|
XM_011525000.1 |
XP_011523302.1
|
9, 10 |
3103 |
|
XM_017024846.1 |
XP_016880335.1
|
9, 10 |
3082 |
|
GenBank mRNA |
AB051525.2 |
- |
8, 9 |
2937 |
|
AF163260.1 |
- |
7, 8 |
1155 |
|
AF220182.1 |
- |
5, 6 |
673 |
|
AK000369.1 |
- |
4, 5 |
444 |
|
AK000758.1 |
- |
8, 9 |
1354 |
|
AK091156.1 |
- |
8, 9 |
2107 |
|
AK091214.1 |
- |
9, 10 |
3035 |
|
AK314316.1 |
- |
4, 5 |
389 |
|
AL832371.1 |
- |
9, 10 |
3018 |
|
AL834251.2 |
- |
9, 10 |
3029 |
|
BC008896.2 |
- |
8, 9 |
2294 |
|
BC033661.1 |
- |
6, 7 |
821 |
|
BC036672.1 |
- |
9, 10 |
3089 |
|
BC040521.1 |
- |
9, 10 |
3043 |
4837394 |
BC040522.1 |
- |
9, 10 |
3091 |
|
BC108279.1 |
- |
7, 8 |
1176 |
|
BX648260.1 |
- |
1, 2 |
1400 |
|
CR627433.1 |
- |
9, 10 |
3111 |
|
View Full Product Details