Gene Transcripts
- Gene Symbol
- CYMP
- Entrez Gene ID
- 643160
- Gene Name
- chymosin pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 1: 110480766 - 110491269 on Build GRCh38
- UniGene ID
- Hs.532720
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003599.2 |
|
2 |
196 |
|
GenBank mRNA |
AK310315.1 |
- |
3 |
312 |
|
View Full Product Details