Gene Transcripts

Gene Symbol
ADAP2
Entrez Gene ID
55803
Gene Name
ArfGAP with dual PH domains 2
Gene Aliases
CENTA2, HSA272195, cent-b
Chromosome Location
Chr. 17: 30921736 - 30959193 on Build GRCh38
UniGene ID
Hs.514063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018404.2 NP_060874.1 11 1818
GenBank mRNA AJ272195.1 - 11 1639
AK074196.1 - 10 1795
BC033758.1 - 11 1632 5214358

View Full Product Details