Gene Transcripts

Gene Symbol
GLRA4
Entrez Gene ID
441509
Gene Name
glycine receptor alpha 4
Gene Aliases
Chromosome Location
Chr. X: 103707344 - 103728624 on Build GRCh38
UniGene ID
Hs.533289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024452.2 NP_001019623.2 6 1016
NM_001172285.1 NP_001165756.1 6 1016
GenBank mRNA AK309010.1 - 6 1016

View Full Product Details