Gene Transcripts

Gene Symbol
USP43
Entrez Gene ID
124739
Gene Name
ubiquitin specific peptidase 43
Gene Aliases
Chromosome Location
Chr. 17: 9643493 - 9729687 on Build GRCh38
UniGene ID
Hs.709621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267576.1 NP_001254505.1 4, 5 934
NM_153210.4 NP_694942.3 4, 5 934
XM_011523640.1 XP_011521942.1 3, 4 654
XM_011523641.1 XP_011521943.1 4, 5 379
XM_011523642.2 XP_011521944.1 4, 5 979
XM_017024159.1 XP_016879648.1 3, 4 875
XM_017024160.1 XP_016879649.1 2, 3 861
XM_017024162.1 XP_016879651.1 4, 5 979
GenBank mRNA AJ583817.2 - 4, 5 841
AK055188.1 - 4, 5 415
BC136368.1 - 4, 5 934 9020355
BC144041.1 - 4, 5 934 9052561
BC171759.1 - 4, 5 934 9054412
BX648526.1 - 2, 3 197

View Full Product Details