Gene Transcripts

Gene Symbol
PSMB11
Entrez Gene ID
122706
Gene Name
proteasome subunit beta 11
Gene Aliases
BETA5T
Chromosome Location
Chr. 14: 23042167 - 23044060 on Build GRCh38
UniGene ID
Hs.508918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099780.1 NP_001093250.1 1 467
GenBank mRNA AB299437.1 - 1 408
AK122797.1 - 1 467

View Full Product Details