Gene Transcripts

Gene Symbol
MYSM1
Entrez Gene ID
114803
Gene Name
Myb like, SWIRM and MPN domains 1
Gene Aliases
2A-DUB, 2ADUB
Chromosome Location
Chr. 1: 58654739 - 58700091 on Build GRCh38
UniGene ID
Hs.744921
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085487.2 NP_001078956.1 8 801
XM_006710314.3 XP_006710377.1 8 817
XM_011540573.2 XP_011538875.1 9 781
XM_011540574.2 XP_011538876.1 8 1049
XM_017000195.1 XP_016855684.1 9 853
XM_017000196.1 XP_016855685.1 6 703
XM_017000197.1 XP_016855686.1 4 481
GenBank mRNA AB067502.1 - 4 827
AK126835.1 - 2 350
BX648168.1 - 5 537

View Full Product Details