Gene Transcripts

Gene Symbol
BIN3
Entrez Gene ID
55909
Gene Name
bridging integrator 3
Gene Aliases
Chromosome Location
Chr. 8: 22620418 - 22669148 on Build GRCh38
UniGene ID
Hs.433722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018688.4 NP_061158.1 7 528
XM_005273573.3 XP_005273630.1 6 434
XM_011544586.2 XP_011542888.1 8 613
GenBank mRNA AF271732.1 - 7 491
AK001289.1 - 5 390
AK023980.1 - 7 475
AK124486.1 - 1 813
AK222470.1 - 7 500
BC001223.1 - 6 412 3534031
BC009824.2 - 7 462 3953604

View Full Product Details