Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018728.3 | NP_061198.2 | 2 | 257 | |
XM_011521781.2 | XP_011520083.1 | 2 | 257 | ||
XM_017022408.1 | XP_016877897.1 | 2 | 257 | ||
XM_017022410.1 | XP_016877899.1 | 2 | 229 | ||
GenBank mRNA | AF272390.1 | - | 2 | 114 | |
AK304136.1 | - | 2 | 228 | ||
BC064841.1 | - | 2 | 203 | ||
BC144611.1 | - | 2 | 129 |