Gene Transcripts
- Gene Symbol
- KRBOX1
- Entrez Gene ID
- 100506243
- Gene Name
- KRAB box domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 42936342 - 42942792 on Build GRCh38
- UniGene ID
- Hs.720172
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001205272.1 |
NP_001192201.1
|
2, 3 |
204 |
|
GenBank mRNA |
AK130974.1 |
- |
2, 3 |
186 |
|
BC062786.1 |
- |
2, 3 |
190 |
|
BC103985.1 |
- |
1, 2 |
249 |
|
BC103986.1 |
- |
1, 2 |
249 |
|
BC103987.1 |
- |
1, 2 |
249 |
|
BC118551.1 |
- |
1, 2 |
249 |
|
BC118662.1 |
- |
1, 2 |
249 |
|
DA207522.1 |
- |
2, 3 |
204 |
|
DB458286.2 |
- |
2, 3 |
186 |
|
- Gene Symbol
- ACKR2
- Entrez Gene ID
- 1238
- Gene Name
- atypical chemokine receptor 2
- Gene Aliases
- CCBP2, CCR10, CCR9, CMKBR9, D6, hD6
- Chromosome Location
- Chr. 3: 42809472 - 42867283 on Build GRCh38
- UniGene ID
- Hs.146346
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DA853159.1 |
- |
3, 4 |
259 |
|
View Full Product Details