Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018948.3 | NP_061821.1 | 4 | 581 | |
XM_005263477.2 | XP_005263534.1 | 4 | 595 | ||
XM_006710697.2 | XP_006710760.1 | 4 | 1765 | ||
XM_011541596.2 | XP_011539898.1 | 3 | 502 | ||
GenBank mRNA | AJ276373.1 | - | 3 | 332 | |
AK096149.1 | - | 4 | 556 | ||
AK315718.1 | - | 4 | 536 | ||
BC025337.1 | - | 4 | 544 | 4810371 |