Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018843.3 | NP_061331.2 | 5 | 600 | |
GenBank mRNA | AF125531.1 | - | 6 | 573 | |
AK223020.1 | - | 5 | 554 | ||
AK290848.1 | - | 5 | 375 | ||
BC027322.1 | - | 4 | 332 | 4345193 | |
BU933121.1 | - | 533 |