Gene Transcripts

Gene Symbol
ERRFI1
Entrez Gene ID
54206
Gene Name
ERBB receptor feedback inhibitor 1
Gene Aliases
GENE-33, MIG-6, MIG6, RALT
Chromosome Location
Chr. 1: 8011719 - 8026333 on Build GRCh38
UniGene ID
Hs.605445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018948.3 NP_061821.1 4 1045
XM_005263477.2 XP_005263534.1 4 1059
XM_006710697.2 XP_006710760.1 4 2229
XM_011541596.2 XP_011539898.1 3 966
GenBank mRNA AJ276373.1 - 3 796
AK027830.1 - 1 140
AK096149.1 - 4 1020
AK315718.1 - 4 1000
BC025337.1 - 4 1008 4810371

View Full Product Details