Gene Transcripts
- Gene Symbol
- LINGO1
- Entrez Gene ID
- 84894
- Gene Name
- leucine rich repeat and Ig domain containing 1
- Gene Aliases
- LERN1, LRRN6A, UNQ201
- Chromosome Location
- Chr. 15: 77613024 - 77820900 on Build GRCh38
- UniGene ID
- Hs.656765
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301186.1 |
NP_001288115.1
|
3, 4 |
244 |
|
NM_001301187.1 |
NP_001288116.1
|
3, 4 |
261 |
|
NM_001301189.1 |
NP_001288118.1
|
3, 4 |
255 |
|
NM_001301191.1 |
NP_001288120.1
|
3, 4 |
440 |
|
NM_001301195.1 |
NP_001288124.1
|
2, 3 |
217 |
|
GenBank mRNA |
BI669707.1 |
- |
3, 4 |
449 |
|
BI825536.1 |
- |
2, 3 |
217 |
|
DA150164.1 |
- |
3, 4 |
244 |
|
DA232085.1 |
- |
3, 4 |
271 |
|
DA483616.1 |
- |
3, 4 |
343 |
|
DA494936.1 |
- |
3, 4 |
377 |
|
DA787289.1 |
- |
3, 4 |
261 |
|
View Full Product Details