Gene Transcripts
- Gene Symbol
- FAM35A
- Entrez Gene ID
- 54537
- Gene Name
- family with sequence similarity 35 member A
- Gene Aliases
- FAM35A1, bA163M19.1
- Chromosome Location
- Chr. 10: 87094478 - 87191468 on Build GRCh38
- UniGene ID
- Hs.500419
- Species
- Homo sapiens
- Gene Symbol
- FAM35BP
- Entrez Gene ID
- 414241
- Gene Name
- family with sequence similarity 35, member A pseudogene
- Gene Aliases
- FAM35B, FAM35EP, bA38L15.1
- Chromosome Location
- Chr. 10: 46610472 - 46651976 on Build GRCh38
- UniGene ID
- Hs.594331
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027632.1 |
|
1 |
168 |
|
- Gene Symbol
- FAM35DP
- Entrez Gene ID
- 439965
- Gene Name
- family with sequence similarity 35, member A pseudogene
- Gene Aliases
- FAM35B2
- Chromosome Location
- Chr. 10: 47689700 - 47731218 on Build GRCh38
- UniGene ID
- Hs.283521
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027634.1 |
|
1 |
168 |
|
View Full Product Details