Gene Transcripts

Gene Symbol
ALG1
Entrez Gene ID
56052
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase
Gene Aliases
CDG1K, HMAT1, HMT-1, HMT1, MT-1, Mat-1, hMat-1
Chromosome Location
Chr. 16: 5071809 - 5087379 on Build GRCh38
UniGene ID
Hs.592086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019109.4 NP_061982.3 3 419
XM_011522565.1 XP_011520867.1 3 226
XM_017023457.1 XP_016878946.1 3 419
XM_017023458.1 XP_016878947.1 3 541
GenBank mRNA AB019038.1 - 3 393
AK075373.1 - 3 409
AK298144.1 - 3 226
AK311475.1 - 3 544
AY359073.1 - 3 403
BC004402.1 - 3 386 3510605
BC031095.1 - 3 398 5276692
BC031873.2 - 4 1145
BM767933.1 - 3 409
DA879477.1 - 3 420

View Full Product Details