Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173516.1 | NP_001166987.1 | 2 | 251 | |
NM_001173517.1 | NP_001166988.1 | 2 | 363 | ||
NM_024597.3 | NP_078873.2 | 2 | 363 | ||
XM_005262472.1 | XP_005262529.1 | 2 | 218 | ||
XM_017029843.1 | XP_016885332.1 | 2 | 215 | ||
GenBank mRNA | AK301478.1 | - | 2 | 252 | |
AL832120.1 | - | 2 | 155 | ||
BC040518.1 | - | 2 | 227 | ||
BC064350.1 | - | 2 | 166 | ||
DB454674.1 | - | 2 | 363 |