Gene Transcripts

Gene Symbol
ALG1
Entrez Gene ID
56052
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase
Gene Aliases
CDG1K, HMAT1, HMT-1, HMT1, MT-1, Mat-1, hMat-1
Chromosome Location
Chr. 16: 5071809 - 5087379 on Build GRCh38
UniGene ID
Hs.592086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019109.4 NP_061982.3 7, 8 913
XM_011522565.1 XP_011520867.1 7, 8 720
XM_017023458.1 XP_016878947.1 7, 8 1035
GenBank mRNA AB019038.1 - 7, 8 887
AK075373.1 - 7, 8 903
AK298144.1 - 7, 8 720
AK311475.1 - 7, 8 1038
AY359073.1 - 7, 8 897
BC004402.1 - 7, 8 880 3510605
BC031095.1 - 7, 8 892 5276692
BC031873.2 - 8, 9 1639

Gene Symbol
TSSC2
Entrez Gene ID
650368
Gene Name
tumor suppressing subtransferable candidate 2 pseudogene
Gene Aliases
Chromosome Location
Chr. 11: 3380961 - 3409148 on Build GRCh38
UniGene ID
Hs.640450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024248.1 5, 6 523
GenBank mRNA AK123648.1 - 6, 7 813
AL528594.3 - 5, 6 513
CN418596.1 - 1, 2 84

View Full Product Details